siRNA Details
Gene Symbol: ECD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135752.1 | NP_001129224.1 | 6, 7 | 1030 | ||
NM_007265.2 | NP_009196.1 | 6, 7 | 1030 | |||
GenBank mRNA | AK022616.1 | - | 6, 7 | 978 | ||
AK315711.1 | - | 6, 7 | 1003 | |||
BC000721.2 | - | 6, 7 | 994 | 3504956 | ||
CR457407.1 | - | 5, 6 | 787 | |||
D88208.1 | - | 6, 7 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
