siRNA Details
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184830.1 | NP_001171759.1 | 3 | 687 | ||
NM_001257403.1 | NP_001244332.1 | 3 | 687 | |||
NM_007268.2 | NP_009199.1 | 3 | 687 | |||
GenBank mRNA | AJ132502.1 | - | 3 | 605 | ||
AK075153.1 | - | 3 | 642 | |||
AK293215.1 | - | 3 | 581 | |||
AK297050.1 | - | 3 | 642 | |||
AL542949.3 | - | 2 | 339 | |||
AL553835.3 | - | 3 | 664 | |||
AY358341.1 | - | 3 | 678 | |||
BC010525.1 | - | 3 | 563 | 4151604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
