siRNA Details
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135746.1 | NP_001129218.1 | 2, 3 | 274 | ||
NM_145043.2 | NP_659480.1 | 2, 3 | 818 | |||
GenBank mRNA | AB079070.1 | - | 1, 2 | 143 | ||
AK056206.1 | - | 2, 3 | 733 | |||
BC013952.2 | - | 2, 3 | 188 | 2964742 | ||
BC013964.2 | - | 2, 3 | 188 | 2964742 | ||
BX537529.1 | - | 2, 3 | 190 | |||
DB452586.1 | - | 2, 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
