siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243962.1 | NP_001230891.1 | 4 | 754 | ||
GenBank mRNA | AK304543.1 | - | 4 | 761 | ||
J00198.1 | - | 3 | 584 | |||
L34095.1 | - | 3 | 579 | |||
L40179.1 | - | 3 | 579 | |||
M81140.1 | - | 4 | 696 | |||
M81141.1 | - | 4 | 710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
