siRNA Details
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135239.1 | NP_001128711.1 | 5 | 752 | ||
NM_001165414.1 | NP_001158886.1 | 6 | 1023 | |||
NM_001165415.1 | NP_001158887.1 | 6 | 926 | |||
NM_001165416.1 | NP_001158888.1 | 6 | 926 | |||
NM_005566.3 | NP_005557.1 | 6 | 926 | |||
NR_028500.1 | 5 | 808 | ||||
GenBank mRNA | AK026515.1 | - | 6 | 752 | ||
AK130035.1 | - | 6 | 755 | |||
AK130105.1 | - | 6 | 750 | |||
AK130587.1 | - | 6 | 753 | |||
AK296089.1 | - | 7 | 671 | |||
AK296667.1 | - | 5 | 579 | |||
AK298834.1 | - | 6 | 877 | |||
AK307477.1 | - | 5 | 634 | |||
AY423727.1 | - | 5 | 685 | |||
BC067223.1 | - | 6 | 756 | 4096518 | ||
CD243986.1 | - | 2 | 228 | |||
CR456775.1 | - | 5 | 654 | |||
CR541714.1 | - | 5 | 654 | |||
EU794632.1 | - | 6 | 751 | |||
X02152.1 | - | 6 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
