siRNA Details
Gene Symbol: NDUFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004545.3 | NP_004536.2 | 3 | 392 | ||
GenBank mRNA | AF050638.1 | - | 3 | 246 | ||
AK055348.1 | - | 3 | 2318 | |||
BC009691.1 | - | 3 | 252 | |||
BC104672.1 | - | 3 | 312 | |||
BM052981.1 | - | 3 | 392 | |||
BX476462.1 | - | 3 | 291 |
