siRNA Details
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 4, 5 | 242 | ||
NM_021603.3 | NP_067614.1 | 4, 5 | 249 | |||
GenBank mRNA | AA309757.1 | - | 4, 5 | 237 | ||
AF241236.1 | - | 4, 5 | 217 | |||
BC005302.1 | - | 4, 5 | 249 | 3828677 | ||
BC013289.1 | - | 4, 5 | 245 | 3686762 | ||
BT006721.1 | - | 4, 5 | 170 | |||
U50743.1 | - | 4, 5 | 206 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 9, 10 | 855 | ||
NM_001243598.2 | NP_001230527.1 | 8, 9 | 793 | |||
GenBank mRNA | CN310658.1 | - | 8, 9 | 428 | ||
FY211982.1 | - | 9, 10 | 452 | |||
FY214031.1 | - | 9, 10 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
