siRNA Details
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 1 | 269 | ||
GenBank mRNA | AY435141.1 | - | 1 | 130 | ||
BC019861.1 | - | 1 | 164 | 4454322 | ||
DB014621.1 | - | 1 | 269 | |||
DQ364250.1 | - | 1 | 138 | |||
J04093.1 | - | 1 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
