siRNA Details
Gene Symbol: NXF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022052.1 | NP_071335.1 | 11 | 1029 | ||
GenBank mRNA | AF346619.1 | - | 11 | 1029 | ||
AJ277527.1 | - | 11 | 1029 | |||
AJ277660.1 | - | 8 | 587 | |||
AK057086.1 | - | 2 | 1191 | |||
AK294826.1 | - | 112 | ||||
BC031616.1 | - | 11 | 1022 | 5170537 |
