siRNA Details
Gene Symbol: LRRIQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079910.1 | NP_001073379.1 | 3 | 307 | ||
XM_011538817.2 | XP_011537119.1 | 3 | 250 | |||
XM_011538818.2 | XP_011537120.1 | 3 | 250 | |||
XM_011538819.2 | XP_011537121.1 | 3 | 250 | |||
XM_011538823.2 | XP_011537125.1 | 3 | 250 | |||
XM_011538824.2 | XP_011537126.1 | 3 | 250 | |||
XM_011538825.2 | XP_011537127.1 | 3 | 250 | |||
XM_017020019.1 | XP_016875508.1 | 3 | 250 | |||
GenBank mRNA | AK022365.1 | - | 3 | 308 | ||
AK307799.1 | - | 3 | 257 | |||
AU122206.2 | - | 3 | 308 | |||
BC093088.1 | - | 3 | 278 | |||
DB447399.2 | - | 3 | 317 |
