siRNA Details
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173473.1 | NP_001166944.1 | 2 | 212 | ||
NM_001173474.1 | NP_001166945.1 | 2 | 297 | |||
NM_004192.3 | NP_004183.2 | 2 | 297 | |||
XM_005274434.3 | XP_005274491.1 | 3 | 403 | |||
GenBank mRNA | AK001090.1 | - | 2 | 193 | ||
AK090498.1 | - | 2 | 249 | |||
AK225770.1 | - | 2 | 219 | |||
AK297805.1 | - | 2 | 195 | |||
AK299231.1 | - | 2 | 215 | |||
AK301844.1 | - | 2 | 206 | |||
BC002508.2 | - | 2 | 183 | 3051194 | ||
BC010089.2 | - | 2 | 181 | 2988425 | ||
DA005122.1 | - | 2 | 297 | |||
DA301876.1 | - | 2 | 252 | |||
DA904467.1 | - | 2 | 212 |
