siRNA Details
Gene Symbol: NOS1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164757.1 | NP_001158229.1 | 5 | 761 | ||
NM_014697.2 | NP_055512.1 | 5 | 776 | |||
GenBank mRNA | AB007933.2 | - | 5 | 776 | ||
BC041382.2 | - | 5 | 778 | |||
BC112295.1 | - | 5 | 477 | 8327763 | ||
BC143771.1 | - | 5 | 462 | 9052285 | ||
BC143780.1 | - | 5 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
