siRNA Details
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260641.3 | XP_005260698.1 | 6 | 2022 | ||
XM_005260643.3 | XP_005260700.1 | 5 | 1689 | |||
XM_005260644.3 | XP_005260701.1 | 4 | 1372 | |||
XM_017027577.1 | XP_016883066.1 | 6 | 2485 | |||
XM_017027581.1 | XP_016883070.1 | 7 | 2330 | |||
XM_017027582.1 | XP_016883071.1 | 7 | 2370 | |||
XM_017027585.1 | XP_016883074.1 | 5 | 2122 | |||
GenBank mRNA | BC025286.1 | - | 4 | 1362 | ||
BC075835.1 | - | 4 | 1362 | |||
BC131747.1 | - | 4 | 1346 | |||
BP268958.1 | - | 1 | 329 | |||
Y10376.1 | - | 6 | 2002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
