siRNA Details
Gene Symbol: CACFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135775.2 | NP_001129247.1 | 3, 4 | 413 | ||
NM_017586.3 | NP_060056.1 | 4, 5 | 539 | |||
XM_011518185.2 | XP_011516487.1 | 4, 5 | 2096 | |||
XM_011518186.1 | XP_011516488.1 | 4, 5 | 592 | |||
GenBank mRNA | AJ011373.1 | - | 4, 5 | 495 | ||
AK074852.1 | - | 4, 5 | 517 | |||
AK075416.1 | - | 3, 4 | 411 | |||
AK075548.1 | - | 4, 5 | 507 | |||
AK304524.1 | - | 5, 6 | 497 | |||
AK311325.1 | - | 4, 5 | 595 | |||
AY358452.1 | - | 4, 5 | 487 | |||
BC030558.1 | - | 4, 5 | 505 | 5223979 | ||
DA699497.1 | - | 3, 4 | 350 | |||
DB165724.1 | - | 4, 5 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
