siRNA Details
Gene Symbol: HOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134670.1 | NP_001128142.1 | 3 | 1647 | ||
NM_138413.3 | NP_612422.2 | 7 | 2136 | |||
GenBank mRNA | BC011916.2 | - | 3 | 1647 | 4310241 | |
BC045550.1 | - | 7 | 2121 | 5274464 | ||
BC057821.1 | - | 3 | 1620 | 5263339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
