siRNA Details
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099225.1 | NP_001092695.1 | 4 | 520 | ||
NM_144609.2 | NP_653210.2 | 5 | 579 | |||
GenBank mRNA | AK056357.1 | - | 5 | 544 | ||
AK098464.1 | - | 1 | 52 | |||
AL833590.1 | - | 4 | 482 | |||
BC023298.1 | - | 5 | 540 | |||
BC047776.2 | - | 5 | 554 | 6023964 | ||
BG706097.1 | - | 4 | 530 | |||
BX648868.1 | - | 4 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
