siRNA Details
Gene Symbol: TCEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080390.3 | NP_525129.1 | 3 | 274 | ||
GenBank mRNA | AF325115.1 | - | 3 | 219 | ||
AK312138.1 | - | 3 | 247 | |||
BC093932.1 | - | 2 | 152 | 7939777 | ||
BC104764.1 | - | 2 | 153 | 8143767 | ||
CD580311.2 | - | 3 | 305 |
