siRNA Details
Gene Symbol: TMEM167A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174909.4 | NP_777569.1 | 4 | 941 | ||
GenBank mRNA | BC028585.1 | - | 4 | 865 | 4836971 | |
BC070122.1 | - | 4 | 859 | |||
BC107575.1 | - | 4 | 852 | 4282548 | ||
BX647979.1 | - | 4 | 841 |
