siRNA Details
Gene Symbol: OR4D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001965.1 | NP_001001965.1 | 1 | 527 | ||
GenBank mRNA | BC136943.1 | - | 1 | 601 | 9020935 | |
BC136945.1 | - | 1 | 601 | 9020937 |
