siRNA Details
Gene Symbol: ATXN7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152749.3 | NP_689962.1 | 3, 4 | 383 | ||
XM_011515954.2 | XP_011514256.1 | 4, 5 | 466 | |||
GenBank mRNA | AK095475.1 | - | 3, 4 | 297 | ||
BC030616.2 | - | 3, 4 | 380 | 4796569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
