siRNA Details
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320340.1 | NP_001307269.1 | 2 | 289 | ||
NM_001320341.1 | NP_001307270.1 | 2 | 180 | |||
NM_001320342.1 | NP_001307271.1 | 2 | 180 | |||
NM_148893.2 | NP_683691.1 | 2 | 180 | |||
NR_135213.1 | 2 | 135 | ||||
GenBank mRNA | AA347778.1 | - | 2 | 131 | ||
AF527534.1 | - | 2 | 154 | |||
AL833119.1 | - | 2 | 153 | |||
BF219509.1 | - | 2 | 136 | |||
BG531443.1 | - | 2 | 113 | |||
BI826588.1 | - | 2 | 288 | |||
DA306370.1 | - | 2 | 129 | |||
DB135755.1 | - | 2 | 183 | |||
DB466086.1 | - | 2 | 116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
