siRNA Details
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 6 | 1460 | ||
NM_015570.3 | NP_056385.1 | 6 | 1460 | |||
XM_011516010.2 | XP_011514312.1 | 6 | 1904 | |||
XM_011516011.2 | XP_011514313.1 | 6 | 1904 | |||
XM_011516012.2 | XP_011514314.1 | 6 | 1904 | |||
XM_011516013.2 | XP_011514315.1 | 6 | 1904 | |||
XM_011516014.2 | XP_011514316.1 | 6 | 1904 | |||
XM_011516017.2 | XP_011514319.1 | 5 | 10458 | |||
XM_011516018.2 | XP_011514320.1 | 6 | 439 | |||
XM_017011951.1 | XP_016867440.1 | 6 | 1904 | |||
GenBank mRNA | AB007902.2 | - | 8 | 1021 | ||
AF326917.1 | - | 6 | 1046 | |||
BC064693.1 | - | 6 | 1468 | 30345831 | ||
JQ670866.1 | - | 6 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
