siRNA Details
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 9, 10 | 2421 | ||
NM_015570.3 | NP_056385.1 | 9, 10 | 2421 | |||
GenBank mRNA | AB007902.2 | - | 11, 12 | 1982 | ||
AF326917.1 | - | 9, 10 | 2007 | |||
BC064693.1 | - | 9, 10 | 2429 | 30345831 | ||
JQ670866.1 | - | 9, 10 | 1686 | |||
JQ670867.1 | - | 1, 2 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
