siRNA Details
Gene Symbol: DIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128226.2 | NP_001121698.1 | 8 | 1354 | ||
NM_001322348.1 | NP_001309277.1 | 7 | 1286 | |||
NM_001322349.1 | NP_001309278.1 | 9 | 1512 | |||
NM_014953.4 | NP_055768.3 | 8 | 1444 | |||
GenBank mRNA | AB001743.1 | - | 8 | 1175 | ||
AB023225.1 | - | 8 | 1151 | |||
AF330044.1 | - | 8 | 1184 | |||
AK025634.1 | - | 6 | 754 | |||
AK314715.1 | - | 8 | 1260 | |||
AL080158.1 | - | 2 | 171 | |||
AL832266.1 | - | 8 | 1237 | |||
BC038101.1 | - | 9 | 1483 | 4827328 | ||
BC056143.1 | - | 8 | 1249 | 5505671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
