siRNA Details
Gene Symbol: DIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322349.1 | NP_001309278.1 | 3 | 695 | ||
NM_014953.4 | NP_055768.3 | 2 | 627 | |||
GenBank mRNA | AB001743.1 | - | 2 | 358 | ||
AF330044.1 | - | 2 | 367 | |||
AK314715.1 | - | 2 | 443 | |||
AL832266.1 | - | 2 | 420 | |||
BC038101.1 | - | 3 | 666 | 4827328 | ||
BC056143.1 | - | 2 | 432 | 5505671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
