siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 7 | 1755 | |||
GenBank mRNA | AB035332.1 | - | 4 | 3534 | ||
AB035333.1 | - | 3 | 3398 | |||
AB035334.1 | - | 5 | 2566 | |||
AB035335.1 | - | 4 | 2111 | |||
AB035337.1 | - | 7 | 1757 | |||
AB035338.1 | - | 7 | 1952 | |||
AB035339.1 | - | 2 | 223 | |||
AF195821.1 | - | 3 | 357 | |||
BX161462.1 | - | 9 | 1796 |
