siRNA Details
Gene Symbol: DIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128226.2 | NP_001121698.1 | 17, 18 | 2547 | ||
NM_001322348.1 | NP_001309277.1 | 16, 17 | 2479 | |||
NM_001322349.1 | NP_001309278.1 | 18, 19 | 2705 | |||
NM_014953.4 | NP_055768.3 | 17, 18 | 2637 | |||
GenBank mRNA | AB001743.1 | - | 17, 18 | 2368 | ||
AB023225.1 | - | 17, 18 | 2344 | |||
AF330044.1 | - | 17, 18 | 2377 | |||
AK001346.1 | - | 1, 2 | 58 | |||
AK025634.1 | - | 15, 16 | 1947 | |||
AK314715.1 | - | 17, 18 | 2453 | |||
AL080158.1 | - | 11, 12 | 1364 | |||
AL832266.1 | - | 17, 18 | 2430 | |||
BC038101.1 | - | 18, 19 | 2676 | 4827328 | ||
BC056143.1 | - | 17, 18 | 2442 | 5505671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
