siRNA Details
Gene Symbol: SND1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027330.1 | 1 | 417 | |||
GenBank mRNA | AF095687.1 | - | 1 | 124 | ||
AF191492.1 | - | 1 | 417 | |||
AK024176.1 | - | 1 | 329 | |||
BC093748.1 | - | 1 | 301 | |||
BC112141.1 | - | 1 | 301 |
