siRNA Details
Gene Symbol: TPRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198479.2 | NP_940881.2 | 2 | 1636 | ||
GenBank mRNA | AK097640.1 | - | 2 | 1636 |
Gene Symbol: TPRXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002223.3 | 3 | 2072 | |||
GenBank mRNA | AK092426.1 | - | 3 | 1997 | ||
BC027729.2 | - | 5 | 2077 |
