siRNA Details
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 3 | 193 | ||
NM_001167741.1 | NP_001161213.1 | 2 | 123 | |||
XM_005248495.3 | XP_005248552.1 | 3 | 195 | |||
XM_017009440.1 | XP_016864929.1 | 2 | 117 | |||
GenBank mRNA | AK091375.1 | - | 1 | 1022 | ||
AK296864.1 | - | 3 | 188 | |||
BC150295.1 | - | 3 | 183 | |||
BX538064.1 | - | 2 | 103 | |||
BX647596.1 | - | 3 | 190 | |||
DB456588.1 | - | 3 | 193 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001162945, AK090249.
Gene Ontology Categories:
