siRNA Details
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014964.4 | NP_055779.2 | 3 | 819 | ||
NM_148921.3 | NP_683723.2 | 3 | 819 | |||
GenBank mRNA | AB028988.1 | - | 3 | 784 | ||
AF062084.1 | - | 1 | 371 | |||
AF062085.1 | - | 1 | 371 | |||
AK001491.1 | - | 1 | 512 | |||
AK001996.1 | - | 1 | 538 | |||
AK024115.1 | - | 3 | 799 | |||
AK298006.1 | - | 2 | 657 | |||
BC070036.1 | - | 3 | 784 | |||
BC093972.1 | - | 1 | 444 | 7939817 | ||
BC093974.1 | - | 1 | 444 | 7939819 | ||
BC143264.1 | - | 1 | 445 | 9051766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
