siRNA Details
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014964.4 | NP_055779.2 | 3 | 890 | ||
NM_148921.3 | NP_683723.2 | 3 | 890 | |||
GenBank mRNA | AB028988.1 | - | 3 | 855 | ||
AF062084.1 | - | 1 | 442 | |||
AF062085.1 | - | 1 | 442 | |||
AK001996.1 | - | 1 | 609 | |||
AK024115.1 | - | 3 | 870 | |||
AK298006.1 | - | 2 | 728 | |||
BC070036.1 | - | 3 | 855 | |||
BC093972.1 | - | 1 | 515 | 7939817 | ||
BC093974.1 | - | 1 | 515 | 7939819 | ||
BC143264.1 | - | 1 | 516 | 9051766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
