siRNA Details
Gene Symbol: C10orf113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010896.2 | NP_001010896.2 | 1 | 131 | ||
NM_001177483.1 | NP_001170954.1 | 1 | 131 | |||
GenBank mRNA | BC140744.1 | - | 1 | 131 | 9021612 | |
BC144447.1 | - | 1 | 131 |
