siRNA Details
Gene Symbol: TAS2R10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023921.1 | NP_076410.1 | 1 | 152 | ||
GenBank mRNA | BC069089.1 | - | 1 | 152 | 7216903 | |
BC101762.1 | - | 1 | 241 | 8069268 | ||
BC101764.1 | - | 1 | 241 | 8069270 |
