siRNA Details
Gene Symbol: CKLF-CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204098.1 | NP_001191027.1 | 1, 2 | 235 | ||
GenBank mRNA | FY211334.1 | - | 1, 2 | 119 |
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181640.2 | NP_857591.1 | 1, 2 | 235 | ||
GenBank mRNA | AF096895.2 | - | 1, 2 | 229 | ||
AK290307.1 | - | 1, 2 | 209 | |||
BC091478.1 | - | 1, 2 | 222 | 30333801 | ||
BU189908.1 | - | 1, 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
