siRNA Details
Gene Symbol: CNTLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017738.3 | NP_060208.2 | 21 | 3537 | ||
XM_005251492.1 | XP_005251549.1 | 21 | 3534 | |||
XM_006716793.3 | XP_006716856.1 | 21 | 3335 | |||
XM_017014839.1 | XP_016870328.1 | 21 | 3537 | |||
XM_017014840.1 | XP_016870329.1 | 21 | 3537 | |||
XM_017014841.1 | XP_016870330.1 | 20 | 3342 | |||
XM_017014844.1 | XP_016870333.1 | 14 | 2213 | |||
XM_017014845.1 | XP_016870334.1 | 12 | 3018 | |||
XM_017014846.1 | XP_016870335.1 | 9 | 1541 | |||
XM_017014847.1 | XP_016870336.1 | 9 | 1534 | |||
GenBank mRNA | AK000283.1 | - | 17 | 2632 | ||
AK021596.1 | - | 9 | 1546 | |||
AK308837.1 | - | 21 | 3489 | |||
BX647069.1 | - | 21 | 3496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
