siRNA Details
Gene Symbol: ANGPTL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018687.6 | NP_061157.3 | 3, 4 | 588 | ||
GenBank mRNA | AF271350.2 | - | 5, 6 | 939 | ||
AY358356.1 | - | 3, 4 | 572 | |||
BC113823.1 | - | 3, 4 | 345 | |||
KF809856.1 | - | 3, 4 | 506 | |||
KF809857.1 | - | 3, 4 | 360 | |||
KF809858.1 | - | 2, 3 | 344 |
