siRNA Details
Gene Symbol: LOC101929512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110593.1 | 3 | 306 | |||
GenBank mRNA | BX107690.1 | - | 3 | 307 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110593.1 | 3 | 306 | |||
GenBank mRNA | BX107690.1 | - | 3 | 307 |