siRNA Details
Gene Symbol: LOC646588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012918.1 | XP_016868407.1 | 4 | 742 | ||
GenBank mRNA | AK057379.1 | - | 4 | 951 | ||
BX648247.1 | - | 1 | 6747 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC646588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012918.1 | XP_016868407.1 | 4 | 742 | ||
GenBank mRNA | AK057379.1 | - | 4 | 951 | ||
BX648247.1 | - | 1 | 6747 |