siRNA Details
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136151.1 | 3 | 663 | |||
GenBank mRNA | CD519625.1 | - | 2 | 461 | ||
DB544287.1 | - | 4 | 405 | |||
DB545731.1 | - | 4 | 417 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136151.1 | 3 | 663 | |||
GenBank mRNA | CD519625.1 | - | 2 | 461 | ||
DB544287.1 | - | 4 | 405 | |||
DB545731.1 | - | 4 | 417 |