siRNA Details
Gene Symbol: PPIAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036506.1 | 1 | 490 | |||
GenBank mRNA | BC157831.1 | - | 1 | 490 | 9057050 | |
BC171823.1 | - | 1 | 490 | 9054477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PPIAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036506.1 | 1 | 490 | |||
GenBank mRNA | BC157831.1 | - | 1 | 490 | 9057050 | |
BC171823.1 | - | 1 | 490 | 9054477 |