siRNA Details
Gene Symbol: LOC105377623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136203.1 | 1 | 250 | |||
GenBank mRNA | CX872222.1 | - | 1 | 218 | ||
DB168916.1 | - | 1 | 251 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105377623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136203.1 | 1 | 250 | |||
GenBank mRNA | CX872222.1 | - | 1 | 218 | ||
DB168916.1 | - | 1 | 251 |