siRNA Details
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012308.2 | NP_036440.1 | 12 | 2062 | ||
NR_027473.1 | 12 | 2062 | ||||
XM_006718480.3 | XP_006718543.1 | 8 | 1080 | |||
XM_011544860.2 | XP_011543162.1 | 12 | 2121 | |||
XM_011544861.1 | XP_011543163.1 | 7 | 774 | |||
XM_017017391.1 | XP_016872880.1 | 12 | 1368 | |||
XM_017017392.1 | XP_016872881.1 | 11 | 1179 | |||
GenBank mRNA | AB023221.2 | - | 12 | 1379 | ||
AK024505.1 | - | 12 | 1304 | |||
BC047371.1 | - | 7 | 774 | 5270744 | ||
BC047486.1 | - | 12 | 2064 | 5296445 | ||
BC064360.1 | - | 12 | 1477 | 5534384 | ||
JQ710741.1 | - | 11 | 1246 | |||
JQ710742.1 | - | 12 | 1368 | |||
JQ710743.1 | - | 11 | 1246 | |||
JQ710744.1 | - | 13 | 1415 |
