siRNA Details
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145204.2 | NP_001138676.2 | 3 | 1258 | ||
XM_005255539.3 | XP_005255596.2 | 3 | 1259 | |||
XM_011522642.2 | XP_011520944.1 | 3 | 1259 | |||
GenBank mRNA | AK299610.1 | - | 4 | 688 |
