siRNA Details
Gene Symbol: LOC730098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320037.1 | NP_001306966.1 | 2 | 110 | ||
NM_001320038.1 | NP_001306967.1 | 1 | 407 | |||
NR_135148.1 | 2 | 110 | ||||
XM_017015106.1 | XP_016870595.1 | 2 | 267 | |||
GenBank mRNA | AK296005.1 | - | 1 | 367 | ||
BC066358.1 | - | 1 | 400 | |||
BM695963.1 | - | 2 | 79 | |||
DB061900.1 | - | 2 | 110 | |||
HY158194.1 | - | 2 | 105 |
