siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011168.2 | - | 15 | 1910 | ||
AB177850.1 | - | 6 | 830 | |||
AK302482.1 | - | 12 | 1684 | |||
AL833267.1 | - | 14 | 1902 | |||
BC036660.1 | - | 13 | 1797 | 5266613 | ||
BC114493.1 | - | 15 | 1892 | 40083182 | ||
BX640787.1 | - | 11 | 2147 | |||
BX648786.1 | - | 14 | 1933 |

More Information
Other Species Targeted

