siRNA Details
Gene Symbol: OR51G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005238.1 | NP_001005238.1 | 1 | 880 | ||
GenBank mRNA | BC136830.1 | - | 1 | 909 | 9020821 | |
BC136831.1 | - | 1 | 909 | 9020822 |
