siRNA Details
Gene Symbol: KDM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015015.2 | NP_055830.1 | 12 | 1878 | ||
XM_005259521.3 | XP_005259578.2 | 13 | 1981 | |||
XM_011527814.2 | XP_011526116.1 | 12 | 1800 | |||
XM_011527816.2 | XP_011526118.1 | 2 | 1330 | |||
XM_011527817.2 | XP_011526119.1 | 5 | 1698 | |||
XM_017026503.1 | XP_016881992.1 | 13 | 1981 | |||
GenBank mRNA | AB020683.2 | - | 12 | 1870 | ||
AK026040.1 | - | 5 | 889 | |||
AK126854.1 | - | 6 | 1418 | |||
AL133622.1 | - | 1 | 254 | |||
BC136611.1 | - | 10 | 1692 | 9020601 | ||
BC144292.1 | - | 11 | 1794 | 9052812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
