siRNA Details
Gene Symbol: KDM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB020683.2 | - | 5 | 565 | ||
BC063889.1 | - | 5 | 554 | |||
BC136611.1 | - | 3 | 387 | 9020601 | ||
BC144292.1 | - | 3 | 387 | 9052812 | ||
DB636535.1 | - | 5 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
