siRNA Details
Gene Symbol: GGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015044.4 | NP_055859.1 | 17 | 1832 | ||
XM_017023074.1 | XP_016878563.1 | 18 | 5521 | |||
XM_017023075.1 | XP_016878564.1 | 17 | 1835 | |||
GenBank mRNA | AB029003.1 | - | 14 | 1462 | ||
AF165531.1 | - | 17 | 1770 | |||
AF190863.1 | - | 17 | 1776 | |||
AF233522.1 | - | 17 | 1775 | |||
AF323754.1 | - | 18 | 1688 | |||
AK024619.1 | - | 17 | 1831 | |||
AK093769.1 | - | 2 | 715 | |||
BC000284.1 | - | 17 | 1814 | 3356490 |
